Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-3-formylbenzeneboronic acid 95%

Supplier: AMBEED, INC
Warning

A170511-1G A170511-250MG A170511-25G A170511-5G
76784-212EA 111 USD InStock
76784-212 76784-214 76784-216 76784-218
4-Benzyloxy-3-formylbenzeneboronic acid 95%
4-Benzyloxy-3-formylbenzeneboronic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₃BO₄
MW: 256.062 g/mol
MDL Number: MFCD02179487
CAS Number: 121124-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty