Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Difluorocyclopropanemethanol 97%

Supplier: AMBEED, INC
Danger

A174269-100MG A174269-250MG
76784-350EA 24 USD InStock
76784-350 76784-352
2,2-Difluorocyclopropanemethanol 97%
2,2-Difluorocyclopropanemethanol
Precaución: for Research Use Only


Formula: C₄H₆F₂O
MDL Number: MFCD06797748
CAS Number: 509072-57-5
UN: 1987
ADR: 3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty