Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-6-piperidinopyrimidine 98%

Supplier: AMBEED, INC
Warning

A178402-1G A178402-250MG
76784-496EA 21.22 USD InStock
76784-496 76784-498 76784-500
4-Amino-6-piperidinopyrimidine 98%
4-Amino-6-piperidinopyrimidine
Precaución: for Research Use Only


Formula: C₉H₁₄N₄
MDL Number: MFCD09999227
CAS Number: 69206-89-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty