Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-trityl-1H-pyrazole 97%

Supplier: AMBEED, INC
Warning

A178466-10G A178466-1G A178466-250MG A178466-5G
76784-508EA 274 USD InStock
76784-508 76784-510 76784-512 76784-514
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-trityl-1H-pyrazole 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-trityl-1H-pyrazole
Precaución: for Research Use Only


Formula: C₂₈H₂₉BN₂O₂
MDL Number: MFCD10698507
CAS Number: 863238-73-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty