Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-5-nitroquinoline 96%

Supplier: AMBEED, INC
Warning

A183222-100MG A183222-250MG
76784-654EA 9 USD InStock
76784-654 76784-656
2,6-Dichloro-5-nitroquinoline 96%
2,6-Dichloro-5-nitroquinoline
Precaución: for Research Use Only


Formula: C₉H₄Cl₂N₂O₂
MDL Number: MFCD12406121
CAS Number: 1209246-34-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty