Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylene bis(4-aminobenzoate) 98%

Supplier: AMBEED, INC
Warning

A186422-100G A186422-25G A186422-500G
76784-738EA 44 USD InStock
76784-738 76784-740 76784-742
Trimethylene bis(4-aminobenzoate) 98%
Trimethylene bis(4-aminobenzoate)
Precaución: for Research Use Only


Formula: C₁₇H₁₈N₂O₄
MDL Number: MFCD00129765
CAS Number: 57609-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty