Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Dimethylamino)propanoic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A187592-100G A187592-10G A187592-1G A187592-25G A187592-5G
76784-792EA 196 USD InStock
76784-792 76784-794 76784-796 76784-798 76784-800
3-(Dimethylamino)propanoic acid hydrochloride 97%
3-(Dimethylamino)propanoic acid hydrochloride
Precaución: for Research Use Only


Formula: C5H12ClNO2
MDL Number: MFCD02093464
CAS Number: 14788-12-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty