Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethynyl-4'-propylbiphenyl 95%

Supplier: AMBEED, INC
Warning

A192031-10G A192031-1G A192031-25G A192031-5G
76784-974EA 54 USD InStock
76784-974 76784-976 76784-978 76784-980 76784-982
4-Ethynyl-4'-propylbiphenyl 95%
4-Ethynyl-4'-propylbiphenyl
Precaución: for Research Use Only


Formula: C₁₇H₁₆
MDL Number: MFCD18072535
CAS Number: 360768-57-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty