Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-6-hydrazinylpyrimidine 97%

Supplier: AMBEED, INC
Warning

A193465-100MG A193465-10G A193465-1G A193465-250MG A193465-25G A193465-5G
76785-014EA 5 USD InStock
76785-014 76785-016 76785-018 76785-020 76785-022 76785-024
4-Hydroxy-6-hydrazinylpyrimidine 97%
4-Hydroxy-6-hydrazinylpyrimidine
Precaución: for Research Use Only


Formula: C₄H₆N₄O
MDL Number: MFCD02695912
CAS Number: 29939-37-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty