Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triethyl-3-phosphonopropionate 95%

Supplier: AMBEED, INC
Warning

A196285-100G A196285-10G A196285-1G A196285-25G A196285-5G
76785-118EA 189 USD InStock
76785-118 76785-120 76785-122 76785-124 76785-126
Triethyl-3-phosphonopropionate 95%
Triethyl-3-phosphonopropionate
Precaución: for Research Use Only


Formula: C₉H₁₉O₅P
MDL Number: MFCD00015167
CAS Number: 3699-67-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty