Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichlorobenzene-1,2-diamine 97%

Supplier: AMBEED, INC
Warning

A197185-10G A197185-1G A197185-250MG A197185-5G
76785-160EA 173 USD InStock
76785-160 76785-162 76785-164 76785-166
3,5-Dichlorobenzene-1,2-diamine 97%
3,5-Dichlorobenzene-1,2-diamine
Precaución: for Research Use Only


Formula: C₆H₆Cl₂N₂
MDL Number: MFCD00628987
CAS Number: 5233-04-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty