Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclohexyl-1H-indole-6-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A197904-100MG A197904-1G A197904-250MG A197904-5G
76785-188EA 35 USD InStock
76785-188 76785-190 76785-192 76785-194
3-Cyclohexyl-1H-indole-6-carboxylic acid 95%
3-Cyclohexyl-1H-indole-6-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₅H₁₇NO₂
MDL Number: MFCD09833286
CAS Number: 494799-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty