Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-(4-Ethylcyclohexyl)benzonitrile 98%

Supplier: AMBEED, INC
Warning

A198952-1G A198952-250MG A198952-25G A198952-5G
76785-228EA 9 USD InStock
76785-228 76785-230 76785-232 76785-234
trans-4-(4-Ethylcyclohexyl)benzonitrile 98%
trans-4-(4-Ethylcyclohexyl)benzonitrile
Precaución: for Research Use Only


Formula: C₁₅H₁₉N
MW: 213.32 g/mol
MDL Number: MFCD09750932
CAS Number: 72928-54-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty