Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dimethylbenzophenone 98%

Supplier: AMBEED, INC
Warning

A199562-100G A199562-25G
76785-262EA 25 USD InStock
76785-262 76785-264
3,4-Dimethylbenzophenone 98%
3,4-Dimethylbenzophenone
Precaución: for Research Use Only


Formula: C₁₅H₁₄O
MW: 210.28 g/mol
MDL Number: MFCD00008525
CAS Number: 2571-39-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty