Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-cyano-2-fluorobenzoate 96%

Supplier: AMBEED, INC
Warning

A201836-1G A201836-250MG A201836-25G A201836-5G
76785-368EA 14 USD InStock
76785-368 76785-370 76785-372 77667-680
Methyl 4-cyano-2-fluorobenzoate 96%
Methyl 4-cyano-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₆FNO₂
MDL Number: MFCD04335187
CAS Number: 175596-01-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty