Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrafluoroisophthalonitrile 98%

Supplier: AMBEED, INC
Danger

A202994-1G A202994-250MG A202994-5G
76785-414EA 11 USD InStock
76785-414 76785-416 76785-418
Tetrafluoroisophthalonitrile 98%
Tetrafluoroisophthalonitrile
Precaución: For research use only.


Formula: C₈F₄N₂
MDL Number: MFCD00013290
CAS Number: 2377-81-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty