Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Topiramate 98%

Supplier: AMBEED, INC
Warning

A206081-100G A206081-500G A206081-5G
76785-508EA 138 USD InStock
76785-508 76785-510 76785-512
Topiramate 98%
Topiramate
Precaución: for Research Use Only


Formula: C₁₂H₂₁NO₈S
MDL Number: MFCD00865320
CAS Number: 97240-79-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty