Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromopyrazine 95%

Supplier: AMBEED, INC
Warning

A207542-10G A207542-1G A207542-25G A207542-5G
76785-566EA 175 USD InStock
76785-566 76785-568 76785-570 76785-572
2,3-Dibromopyrazine 95%
2,3-Dibromopyrazine
Precaución: for Research Use Only


Formula: C₄H₂Br₂N₂
MDL Number: MFCD02684897
CAS Number: 95538-03-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty