Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl methyl(piperidin-4-ylmethyl)carbamate 95%

Supplier: AMBEED, INC
Warning

A209988-1G A209988-250MG A209988-5G
76785-650EA 118 USD InStock
76785-650 76785-652 76785-654
tert-Butyl methyl(piperidin-4-ylmethyl)carbamate 95%
tert-Butyl methyl(piperidin-4-ylmethyl)carbamate
Precaución: for Research Use Only


Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD06800387
CAS Number: 138022-04-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty