Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-methylthiopyrimidine 97%

Supplier: AMBEED, INC
Warning

A212738-100MG A212738-10G A212738-1G A212738-250MG A212738-5G
76785-726EA 7 USD InStock
76785-726 76785-728 76785-730 76785-732 76785-734
4-Chloro-6-methylthiopyrimidine 97%
4-Chloro-6-methylthiopyrimidine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂S
MDL Number: MFCD09870036
CAS Number: 89283-48-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty