Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetramethyl methylenebisphosphonate 98%

Supplier: AMBEED, INC
Warning

A213648-1G A213648-250MG A213648-25G A213648-5G
76785-746EA 20 USD InStock
76785-746 76785-748 76785-750 76785-752
Tetramethyl methylenebisphosphonate 98%
Tetramethyl methylenebisphosphonate
Precaución: for Research Use Only


Formula: C₅H₁₄O₆P₂
MDL Number: MFCD00014884
CAS Number: 16001-93-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty