Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tiagabine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A214929-100MG A214929-10MG A214929-1MG A214929-250MG A214929-50MG
76785-786EA 162 USD InStock
76785-786 76785-788 76785-790 76785-792 76785-794
Tiagabine hydrochloride 98%
Tiagabine hydrochloride
Precaución: for Research Use Only


Formula: C₂₀H₂₅NO₂S₂
MDL Number: MFCD07369025
CAS Number: 145821-59-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty