Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diazaspiro[3.4]octane-2-carboxylic acid tert-butylester 97%

Supplier: AMBEED, INC
Warning

A216997-100MG A216997-10G A216997-1G A216997-250MG A216997-5G
76785-834EA 9 USD InStock
76785-834 76785-836 76785-838 76785-840 76785-842
2,6-Diazaspiro[3.4]octane-2-carboxylic acid tert-butylester 97%
2,6-Diazaspiro[3.4]octane-2-carboxylic acid tert-butylester
Precaución: for Research Use Only


Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD08234737
CAS Number: 885270-84-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty