Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2,6-difluorophenol hydrochloride 98%

Supplier: AMBEED, INC
Warning

A217145-100MG A217145-1G A217145-250MG
76785-844EA 18 USD InStock
76785-844 76785-846 76785-848
4-Amino-2,6-difluorophenol hydrochloride 98%
4-Amino-2,6-difluorophenol hydrochloride
Precaución: for Research Use Only


Formula: C₆H₆ClF₂NO
MW: 181.5678 g/mol
MDL Number: MFCD12197391
CAS Number: 220353-22-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty