Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1-methyl-1H-pyrazole 97%

Supplier: AMBEED, INC
Warning

A221966-10G A221966-250MG A221966-25G A221966-5G
76786-020EA 67 USD InStock
76786-020 76786-022 76786-024 76786-026
5-Bromo-1-methyl-1H-pyrazole 97%
5-Bromo-1-methyl-1H-pyrazole
Precaución: For research use only.


Formula: C₄H₅BrN₂
MDL Number: MFCD11656572
CAS Number: 361476-01-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty