Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Piperidine-3-carboxylic acid hydrochloride 98%

Supplier: AMBEED, INC
Warning

A225802-10G A225802-1G A225802-5G
76786-116EA 57 USD InStock
76786-116 76786-118 76786-120
(S)-Piperidine-3-carboxylic acid hydrochloride 98%
(S)-Piperidine-3-carboxylic acid hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₂ClNO₂
MDL Number: MFCD00211281
CAS Number: 851956-01-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty