Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulfathiazole 98+%

Supplier: AMBEED, INC
Warning

A227334-1000G A227334-100G A227334-25G A227334-500G
76786-146EA 183 USD InStock
76786-146 76786-148 76786-150 76786-152
Sulfathiazole 98+%
Sulfathiazole
Precaución: For research use only.


Formula: C₉H₉N₃O₂S₂
MDL Number: MFCD00005319
CAS Number: 72-14-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty