Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rhodium (III) acetylacetonate 98%

Supplier: AMBEED, INC
Danger

A228348-100MG A228348-1G A228348-250MG
76786-178EA 35 USD InStock
76786-178 76786-180 76786-182
Rhodium (III) acetylacetonate 98%
Rhodium (III) acetylacetonate
Precaución: for Research Use Only


Formula: C₁₅H₂₁O₆Rh
MW: 400.23
MDL Number: MFCD00083144
CAS Number: 14284-92-5
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty