Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodo-4'-propylbiphenyl 98%

Supplier: AMBEED, INC
Warning

A232950-100MG A232950-1G A232950-250MG A232950-5G
76786-322EA 13 USD InStock
76786-322 76786-324 76786-326 76786-328
4-Iodo-4'-propylbiphenyl 98%
4-Iodo-4'-propylbiphenyl
Precaución: for Research Use Only


Formula: C₁₅H₁₅I
MDL Number: MFCD12923241
CAS Number: 782477-81-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty