Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Hydroxy-D-proline hydrochloride 97%

Supplier: AMBEED, INC
Warning

A236256-10G A236256-1G A236256-5G A236256-25G
76786-404EA 75 USD InStock
76786-404 76786-406 76786-408 77667-946
trans-4-Hydroxy-D-proline hydrochloride 97%
trans-4-Hydroxy-D-proline hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₀ClNO₃
MDL Number: MFCD08436165
CAS Number: 142347-81-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty