Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl ((benzyloxy)carbonyl)-L-tyrosinate 97%

Supplier: AMBEED, INC
Warning

A239029-10G A239029-1G A239029-25G A239029-5G
76786-458EA 26 USD InStock
76786-458 76786-460 76786-462 76786-464
tert-Butyl ((benzyloxy)carbonyl)-L-tyrosinate 97%
tert-Butyl ((benzyloxy)carbonyl)-L-tyrosinate
Precaución: for Research Use Only


Formula: C₂₁H₂₅NO₅
MW: 371.42 g/mol
MDL Number: MFCD00191166
CAS Number: 16881-33-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty