Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodobenzo[d]thiazole 95%

Supplier: AMBEED, INC
Warning

A243341-10G A243341-1G A243341-250MG A243341-25G A243341-5G
76786-600EA 230 USD InStock
76786-600 76786-602 76786-604 76786-606 76786-608
2-Iodobenzo[d]thiazole 95%
2-Iodobenzo[d]thiazole
Precaución: for Research Use Only


Formula: C₇H₄INS
MDL Number: MFCD00971971
CAS Number: 1123-99-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty