Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(((Benzyloxy)carbonyl)amino)-5-ethoxy-5-oxopentanoic acid 97%

Supplier: AMBEED, INC
Warning

A245772-10G A245772-1G A245772-250MG A245772-5G
76786-672EA 157 USD InStock
76786-672 76786-674 76786-676 76786-678
(S)-2-(((Benzyloxy)carbonyl)amino)-5-ethoxy-5-oxopentanoic acid 97%
(S)-2-(((Benzyloxy)carbonyl)amino)-5-ethoxy-5-oxopentanoic acid
Precaución: for Research Use Only


Formula: C₁₅H₁₉NO₆
MDL Number: MFCD00274757
CAS Number: 35726-62-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty