Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-(S)-3-amino-4-(4-chlorophenyl)butyric acid 97%

Supplier: AMBEED, INC
Warning

A248294-100MG A248294-1G A248294-250MG
76786-740EA 10 USD InStock
76786-740 76786-742 76786-744
Boc-(S)-3-amino-4-(4-chlorophenyl)butyric acid 97%
Boc-(S)-3-amino-4-(4-chlorophenyl)butyric acid
Precaución: for Research Use Only


Formula: C₁₅H₂₀ClNO₄
MDL Number: MFCD01318372
CAS Number: 270596-42-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty