Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pneumocandin B0 98%

Supplier: AMBEED, INC
Warning

A248329-10MG A248329-1G A248329-1MG A248329-5MG A248329-100MG
76786-746EA 15 USD InStock
76786-746 76786-748 76786-750 76786-752 77668-028
Pneumocandin B0 98%
Pneumocandin B0
Precaución: for Research Use Only


Formula: C₅₀H₈₀N₈O₁₇
MW: 1065.23 g/mol
MDL Number: MFCD28168025
CAS Number: 135575-42-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty