Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminomethylpyridazine 95%

Supplier: AMBEED, INC
Warning

A248559-100MG A248559-250MG A248559-50MG
76786-764EA 179 USD InStock
76786-764 76786-766 76786-768
3-Aminomethylpyridazine 95%
3-Aminomethylpyridazine
Precaución: for Research Use Only


Formula: C₅H₇N₃
MDL Number: MFCD05664656
CAS Number: 93319-65-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty