Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Aminomethyl)-2-methoxyphenol 97%

Supplier: AMBEED, INC
Danger

A250193-10G A250193-1G A250193-250MG A250193-5G
76786-804EA 166 USD InStock
76786-804 76786-806 76786-808 77601-018
4-(Aminomethyl)-2-methoxyphenol 97%
4-(Aminomethyl)-2-methoxyphenol
Precaución: for Research Use Only


Formula: C₈H₁₁NO₂
MDL Number: MFCD00044577
CAS Number: 1196-92-5
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty