Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrolo[3,2-b]pyridin-5(4H)-one 95%

Supplier: AMBEED, INC
Warning

A258803-100MG A258803-1G A258803-250MG
76786-928EA 15 USD InStock
76786-928 76786-930 76786-932
1H-Pyrrolo[3,2-b]pyridin-5(4H)-one 95%
1H-Pyrrolo[3,2-b]pyridin-5(4H)-one
Precaución: for Research Use Only


Formula: C₇H₆N₂O
MW: 134.13 g/mol
MDL Number: MFCD13175849
CAS Number: 17322-91-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty