Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Acetyl-L-tryptophan ethyl ester 98%

Supplier: AMBEED, INC
Warning

A265487-1G A265487-250MG A265487-25G A265487-5G
76787-078EA 26 USD InStock
76787-078 76787-080 76787-082 76787-084
N-Acetyl-L-tryptophan ethyl ester 98%
N-Acetyl-L-tryptophan ethyl ester
Precaución: for Research Use Only


Formula: C₁₅H₁₈N₂O₃
MDL Number: MFCD00005643
CAS Number: 2382-80-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty