Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-1-methyl-1H-indazole 97%

Supplier: AMBEED, INC
Warning

A267315-1G A267315-250MG A267315-5G
76787-112EA 12 USD InStock
76787-112 76787-114 76787-116
6-Bromo-1-methyl-1H-indazole 97%
6-Bromo-1-methyl-1H-indazole
Precaución: for Research Use Only


Formula: C₈H₇BrN₂
MDL Number: MFCD09878569
CAS Number: 590417-94-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty