Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Bromomethyl)-4-phenylthiazole 95%

Supplier: AMBEED, INC
Danger

A273843-100MG A273843-1G A273843-250MG
76787-240EA 70 USD InStock
76787-240 76787-242 76787-244
2-(Bromomethyl)-4-phenylthiazole 95%
2-(Bromomethyl)-4-phenylthiazole
Precaución: for Research Use Only


Formula: C₁₀H₈BrNS
MDL Number: MFCD12405586
CAS Number: 78502-79-1
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty