Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-bromophenol 98%

Supplier: AMBEED, INC
Warning

A276353-100G A276353-10G A276353-1G A276353-250MG A276353-25G A276353-5G
76787-286EA 396 USD InStock
76787-286 76787-288 76787-290 76787-292 76787-294 76787-296
4-Amino-2-bromophenol 98%
4-Amino-2-bromophenol
Precaución: for Research Use Only


Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD06656565
CAS Number: 16750-67-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty