Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloropyrazine-2-carboxylic acid hydrazide 98%

Supplier: AMBEED, INC
Warning

A276788-1G A276788-250MG
76787-312EA 192 USD InStock
76787-312 76787-314
5-Chloropyrazine-2-carboxylic acid hydrazide 98%
5-Chloropyrazine-2-carboxylic acid hydrazide
Precaución: for Research Use Only


Formula: C5H5ClN4O
MDL Number: MFCD12964049
CAS Number: 848952-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty