Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-6-methoxyindole-2-boronic acid 98%

Supplier: AMBEED, INC
Warning

A279831-1G A279831-250MG A279831-100MG
76787-372EA 40 USD InStock
76787-372 76787-374 77668-240
1-Boc-6-methoxyindole-2-boronic acid 98%
1-Boc-6-methoxyindole-2-boronic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₈BNO₅
MDL Number: MFCD06659830
CAS Number: 850568-65-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty