Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Oxa-8-azaspiro[4.5]decane hydrochloride 97%

Supplier: AMBEED, INC
Warning

A284640-100MG A284640-250MG A284640-5G
76787-440EA 12 USD InStock
76787-440 76787-442 76787-444
1-Oxa-8-azaspiro[4.5]decane hydrochloride 97%
1-Oxa-8-azaspiro[4.5]decane hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₆ClNO
MDL Number: MFCD11042294
CAS Number: 3970-79-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty