Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Diiodopyrimidine 95%

Supplier: AMBEED, INC
Warning

A286103-1G A286103-250MG
76787-460EA 35 USD InStock
76787-460 76787-462
2,4-Diiodopyrimidine 95%
2,4-Diiodopyrimidine
Precaución: for Research Use Only


Formula: C₄H₂I₂N₂
MDL Number: MFCD09743716
CAS Number: 262353-34-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty