Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Dimethylamino)benzophenone 98%

Supplier: AMBEED, INC
Warning

A289191-100G A289191-1G A289191-25G A289191-5G
76787-510EA 395 USD InStock
76787-510 76787-512 76787-514 76787-516
4-(Dimethylamino)benzophenone 98%
4-(Dimethylamino)benzophenone
Precaución: for Research Use Only


Formula: C₁₅H₁₅NO
MW: 225.29 g/mol
MDL Number: MFCD00008311
CAS Number: 530-44-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty