Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3,6-dibromopyrazine-2-carboxylate 97%

Supplier: AMBEED, INC
Warning

A290056-10G A290056-1G A290056-250MG A290056-25G A290056-5G
76787-522EA 35 USD InStock
76787-522 76787-524 76787-526 76787-528 76787-530
Methyl-3,6-dibromopyrazine-2-carboxylate 97%
Methyl-3,6-dibromopyrazine-2-carboxylate
Precaución: for Research Use Only


Formula: C₆H₄Br₂N₂O₂
MDL Number: MFCD11976863
CAS Number: 13301-04-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty