Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Scutellarein 98%

Supplier: AMBEED, INC
Warning

A295800-100MG A295800-50MG A295800-250MG
76787-624EA 95 USD InStock
76787-624 76787-626 76787-628 76787-630 76787-632 77668-320
Scutellarein 98%
Scutellarein
Precaución: for Research Use Only


Formula: C₁₅H₁₀O₆
MDL Number: MFCD00017692
CAS Number: 529-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty