Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Iodo-N-phenylpicolinamide 95+%

Supplier: AMBEED, INC
Warning

A295998-1G A295998-250MG A295998-5G
76787-634EA 64 USD InStock
76787-634 76787-636 76787-638
3-Iodo-N-phenylpicolinamide 95+%
3-Iodo-N-phenylpicolinamide
Precaución: for Research Use Only


Formula: C12H9IN2O
MDL Number: MFCD11109824
CAS Number: 57841-90-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty